5-Chloro-2-ethynylpyridine hydrochloride 95%

Supplier: Ambeed
Warning

A147786-100MG A147786-1G A147786-250MG
76984-522EA 42 USD
76984-522 76984-524 76984-526
5-Chloro-2-ethynylpyridine hydrochloride 95%
5-Chloro-2-ethynylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₅Cl₂N
MDL Number: MFCD26743576
CAS Number: 1956341-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR