N-Methylpropionamide 99%

Supplier: Ambeed
Warning

A147740-1G A147740-25G
76997-192EA 8.49 USD
76997-192 76997-194
N-Methylpropionamide 99%
N-Methylpropionamide

Precaución: for Research Use Only

Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD00009300
CAS Number: 1187-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR