Fmoc-D-Dap(Alloc)-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((allyloxy)carbonyl)amino)propanoic acid
A147660-1G
A147660-250MG
A147660-5G
76984-514EA
36
USD
76984-514
76984-516
77889-969
Fmoc-D-Dap(Alloc)-OH 98%
Fmoc-D-Dap(Alloc)-OH
Precaución: for Research Use Only
Formula:
C₂₂H₂₂N₂O₆ MW: 410.5 g/mol |
MDL Number:
MFCD01076137 CAS Number: 178924-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...