3-Fluoro-5-iodophenol 96%

Supplier: Ambeed
Danger

A147607-100MG A147607-250MG
76984-508EA 41 USD
76984-508 76984-510
3-Fluoro-5-iodophenol 96%
3-Fluoro-5-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄FIO
MDL Number: MFCD16999911
CAS Number: 939771-60-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR