4-Methoxy-2-oxo-1,2-dihydropyridine-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A147498-100MG
A147498-1G
A147498-250MG
77010-612EA
27.59
USD
77010-612
77010-614
77010-616
4-Methoxy-2-oxo-1,2-dihydropyridine-3-carboxylic acid 97%
4-Methoxy-2-oxo-1,2-dihydropyridine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₄ MW: 169.13 g/mol |
MDL Number:
MFCD20654516 CAS Number: 87853-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...