1-Pyridin-4-ylpiperidin-4-one 97%

Supplier: Ambeed
Warning

A147419-100MG A147419-1G A147419-250MG A147419-5G
76997-184EA 23 USD
76997-184 76997-186 76997-188 76997-190
1-Pyridin-4-ylpiperidin-4-one 97%
1-Pyridin-4-ylpiperidin-4-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD09030756
CAS Number: 126832-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR