5-Bromo-7-nitro-1H-indazole 95%

Supplier: Ambeed
Warning

A147412-100MG A147412-1G A147412-250MG A147412-25G A147412-5G
76997-174EA 14.86 USD
76997-174 76997-176 76997-178 76997-180 76997-182
5-Bromo-7-nitro-1H-indazole 95%
5-Bromo-7-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD03617604
CAS Number: 316810-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR