1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride 98%

Supplier: Ambeed
Danger

A147356-1G A147356-250MG A147356-5G
76997-168EA 18 USD
76997-168 76997-170 76997-172
1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride 98%
1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride

Precaución: for Research Use Only

Formula: C₇H₉ClFNO
MDL Number: MFCD01114584
CAS Number: 215599-91-0
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR