3,4-Difluoro-5-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A147262-10G A147262-1G A147262-250MG A147262-5G
77010-588EA 217 USD
77010-588 77010-590 77010-592 77010-594
3,4-Difluoro-5-methoxybenzaldehyde 95%
3,4-Difluoro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD04972450
CAS Number: 881190-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR