(1-Naphthylmethyl)triphenylphosphonium chloride 95%

Supplier: Ambeed
Warning

Synonyms: (Naphthalen-1-ylmethyl)triphenylphosphonium chloride

A147255-1G A147255-25G A147255-5G
76997-162EA 18.04 USD
76997-162 76997-164 76997-166
(1-Naphthylmethyl)triphenylphosphonium chloride 95%
(1-Naphthylmethyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₉H₂₄ClP
MW: 438.9276 g/mol
MDL Number: MFCD00031658
CAS Number: 23277-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR