(2R,4S)-4-Fluoropyrrolidine-2-carboxylic acid, N-BOC protected 95%

Supplier: Ambeed
Warning

A147188-100MG A147188-250MG A147188-1G A147188-5G A147188-10G
76984-498EA 10 USD
76984-498 76984-500 77592-756 77592-758 77889-968
(2R,4S)-4-Fluoropyrrolidine-2-carboxylic acid, N-BOC protected 95%
(2R,4S)-4-Fluoropyrrolidine-2-carboxylic acid, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₀H₁₆FNO₄
MDL Number: MFCD08706411
CAS Number: 681128-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR