2-Amino-4-(3-bromophenyl)pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Bromophenyl)pyrimidin-2-amine

A147045-1G A147045-250MG
76984-490EA 145.35 USD
76984-490 76984-492
2-Amino-4-(3-bromophenyl)pyrimidine 98%
2-Amino-4-(3-bromophenyl)pyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₈BrN₃
MW: 250.10 g/mol
MDL Number: MFCD01940446
CAS Number: 392307-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR