Fmoc-3,4-dichloro-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(3,4-DiCl)-OH, (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4-dichlorophenyl)propanoic acid, 3,4-Dichloro-L-phenylalanine, N-FMOC protected

A147034-1G A147034-250MG A147034-5G A147034-25G
77027-934EA 65 USD
77027-934 77027-936 77027-938 77871-717
Fmoc-3,4-dichloro-L-phenylalanine 95%
Fmoc-3,4-dichloro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₁₉Cl₂NO₄
MW: 456.33 g/mol
MDL Number: MFCD00273473
CAS Number: 177966-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR