4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one 97%
Supplier: Ambeed
|
Danger
|
A146976-100G
A146976-10G
A146976-500G
A146976-5G
A146976-25G
77010-574EA
24
USD
77010-574
77010-576
77010-578
77010-580
77317-180
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one 97%
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one
Precaución: for Research Use Only
Formula:
C₅H₅ClO₃ |
MDL Number:
MFCD07787494 CAS Number: 80841-78-7 UN: 3265 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...