Methyl 3-hydroxy-2-nitrobenzoate 95%

Supplier: Ambeed
Warning

A146902-1G A146902-250MG A146902-5G A146902-25G
76997-134EA 33 USD
76997-134 76997-136 76997-138 77881-144
Methyl 3-hydroxy-2-nitrobenzoate 95%
Methyl 3-hydroxy-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MDL Number: MFCD09951956
CAS Number: 89942-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR