Z-Lys(Boc)-OH·DCHA 97%

Supplier: Ambeed
Warning

Synonyms: Cbz-Lys(Boc)-OH·DCHA, Z-Lys(Boc)-OH·DCHA, Z-Lys(Boc)-OH.DCHA

A146901-25G A146901-5G
76997-130EA 39 USD
76997-130 76997-132
Z-Lys(Boc)-OH·DCHA 97%
Z-Lys(Boc)-OH·DCHA

Precaución: for Research Use Only

Formula: C₃₁H₅₁N₃O₆
MW: 561.764 g/mol
MDL Number: MFCD00038269
CAS Number: 2212-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR