3,4-Dihydroxy-5-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A146884-100G A146884-10G A146884-25G A146884-500G A146884-5G
76997-118EA 24 USD
76997-118 76997-120 76997-122 76997-124 76997-126 76997-128
3,4-Dihydroxy-5-nitrobenzaldehyde 97%
3,4-Dihydroxy-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅NO₅
MDL Number: MFCD00871897
CAS Number: 116313-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR