4-(Bromomethyl)phenyl acetate 97%, Ambeed.Inc

Supplier: Ambeed
Danger

A146596-5G A146596-1G A146596-250MG
77010-566EA 375.56 USD
77010-566 77566-990 77683-251
4-(Bromomethyl)phenyl acetate 97%, Ambeed.Inc
4-(Bromomethyl)phenyl acetate

Precaución: For research use only.

Formula: C₉H₉BrO₂
MW: 229.071 g/mol
MDL Number: MFCD17013576
CAS Number: 52727-95-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR