Ethyl 4-(2-hydroxyphenyl)butanoate 95%
Supplier: Ambeed
|
Warning
|
A146574-100MG
A146574-1G
A146574-250MG
77027-924EA
37.14
USD
77027-924
77027-926
77027-928
Ethyl 4-(2-hydroxyphenyl)butanoate 95%
Ethyl 4-(2-hydroxyphenyl)butanoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₆O₃ MW: 208.25 g/mol |
MDL Number:
MFCD12756195 CAS Number: 20921-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...