5-(Benzyloxy)-2-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A146328-100MG A146328-1G
76984-474EA 79.57 USD
76984-474 76984-476
5-(Benzyloxy)-2-hydroxybenzaldehyde 95%
5-(Benzyloxy)-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MW: 228.24 g/mol
MDL Number: MFCD00436292
CAS Number: 56979-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR