2-Iodo-6-methylpyridin-3-amine 98%

Supplier: Ambeed
Warning

A146326-100MG A146326-1G A146326-250MG
77010-548EA 37 USD
77010-548 77010-550 77010-552
2-Iodo-6-methylpyridin-3-amine 98%
2-Iodo-6-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MDL Number: MFCD11109566
CAS Number: 1211596-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR