5-Cyano-2,3-dihydro-1H-indole 97%

Supplier: Ambeed
Warning

Synonyms: Indoline-5-carbonitrile

A146134-100MG A146134-1G A146134-250MG
77010-528EA 39.26 USD
77010-528 77010-530 77010-532
5-Cyano-2,3-dihydro-1H-indole 97%
5-Cyano-2,3-dihydro-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.173 g/mol
MDL Number: MFCD07371626
CAS Number: 15861-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR