Methyl trans-3-hydroxycyclobutanecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: trans-Methyl 3-hydroxycyclobutanecarboxylate

A146126-100MG A146126-1G A146126-250MG A146126-25G A146126-5G
76984-462EA 42.44 USD
76984-462 76984-464 76984-466 76984-468 76984-470
Methyl trans-3-hydroxycyclobutanecarboxylate 95%
Methyl trans-3-hydroxycyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.142 g/mol
MDL Number: MFCD18483114
CAS Number: 63485-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR