2-Amino-5-iodo-3-nitropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Iodo-3-nitropyridin-2-amine

A146027-1G A146027-25G A146027-5G
77010-524EA 10 USD
77010-524 77010-526 77889-965
2-Amino-5-iodo-3-nitropyridine 97%
2-Amino-5-iodo-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄IN₃O₂
MW: 265.009 g/mol
MDL Number: MFCD04038464
CAS Number: 25391-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR