5-Amino-2,3-difluorobenzonitrile 97%

Supplier: Ambeed
Warning

A145929-1G A145929-250MG A145929-100MG
76984-458EA 109 USD
76984-458 76984-460 77889-964
5-Amino-2,3-difluorobenzonitrile 97%
5-Amino-2,3-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂
MDL Number: MFCD20726994
CAS Number: 1247885-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR