8-Chloro-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

A145876-1G A145876-250MG A145876-100MG
77010-520EA 461.5 USD
77010-520 77010-522 77683-249
8-Chloro-1,2,3,4-tetrahydroquinoline 95%
8-Chloro-1,2,3,4-tetrahydroquinoline

Precaución: For research use only.

Formula: C₉H₁₀ClN
MW: 167.64 g/mol
MDL Number: MFCD09034968
CAS Number: 90562-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR