2-Methoxy-6-nitrobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A145842-1G
A145842-250MG
A145842-5G
77027-890EA
35
USD
77027-890
77027-892
77027-894
2-Methoxy-6-nitrobenzoic acid 97%
2-Methoxy-6-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇NO₅ |
MDL Number:
MFCD00091878 CAS Number: 53967-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...