9-Fluorenone oxime 97%

Supplier: Ambeed
Danger

Synonyms: 9H-Fluoren-9-one oxime

A145705-1G A145705-25G A145705-5G
76997-072EA 10 USD
76997-072 76997-074 76997-076
9-Fluorenone oxime 97%
9-Fluorenone oxime

Precaución: for Research Use Only

Formula: C₁₃H₉NO
MDL Number: MFCD00016356
CAS Number: 2157-52-0
UN: 2926
ADR: 4.1,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR