Methyl-4-fluoro-1H-indole-2-carboxylate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Methyl 4-fluoro-1H-indole-2-carboxylate

A145694-100MG A145694-1G A145694-250MG A145694-5G
77010-514EA 5 USD
77010-514 77010-516 77010-518 77683-248
Methyl-4-fluoro-1H-indole-2-carboxylate 95%, Ambeed.Inc
Methyl-4-fluoro-1H-indole-2-carboxylate

Precaución: For research use only.

Formula: C₁₀H₈FNO₂
MW: 193.18 g/mol
MDL Number: MFCD06653401
CAS Number: 113162-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR