4-Bromo-3-fluoroaniline, N-CBZ protected 97%

Supplier: Ambeed
Warning

A145669-100MG A145669-1G A145669-250MG A145669-5G
76984-450EA 8 USD
76984-450 76984-452 76984-454 76984-456
4-Bromo-3-fluoroaniline, N-CBZ protected 97%
4-Bromo-3-fluoroaniline, N-CBZ protected

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrFNO₂
MDL Number: MFCD23381075
CAS Number: 510729-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR