3-Oxo-1-phenyl-2,7,10-trioxa-4-azadodecan-12-oic acid 95%

Supplier: Ambeed
Warning

Synonyms: Cbz-NH-PEG2-CH2COOH

A145646-100MG A145646-1MG A145646-250MG A145646-5G A145646-1G A145646-25G
76984-442EA 15.92 USD
76984-442 76984-444 76984-446 76984-448 77611-346 77683-247
3-Oxo-1-phenyl-2,7,10-trioxa-4-azadodecan-12-oic acid 95%
3-Oxo-1-phenyl-2,7,10-trioxa-4-azadodecan-12-oic acid

Precaución: For research use only.

Formula: C₁₄H₁₉NO₆
MW: 297.30 g/mol
MDL Number: MFCD23379879
CAS Number: 165454-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR