Imidazo[1,2-a]pyrimidine 97%

Supplier: Ambeed
Warning

A145614-100G A145614-10G A145614-25G A145614-5G A145614-1G A145614-250MG
76997-050EA 778 USD
76997-050 76997-052 76997-054 77600-090 77671-066 77881-138
Imidazo[1,2-a]pyrimidine 97%
Imidazo[1,2-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₅N₃
MDL Number: MFCD06245377
CAS Number: 274-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR