1-Methyl-1H-benzimidazol-6-amine 98%

Supplier: Ambeed
Warning

A145567-100MG A145567-10G A145567-1G A145567-250MG A145567-5G
77027-872EA 26 USD
77027-872 77027-874 77027-876 77027-878 77027-880
1-Methyl-1H-benzimidazol-6-amine 98%
1-Methyl-1H-benzimidazol-6-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MDL Number: MFCD09764050
CAS Number: 26530-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR