3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 3-hydroxy-1H-pyrazole-4-carboxylate
A145549-100MG
A145549-10G
A145549-1G
A145549-250MG
A145549-5G
77010-504EA
10.61
USD
77010-504
77010-506
77010-508
77010-510
77010-512
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 98%
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₃ MW: 156.14 g/mol |
MDL Number:
MFCD12131138 CAS Number: 7251-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...