2-(4-Methoxy-benzyl)-2H-pyrazol-3-ylamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Methoxybenzyl)-1H-pyrazol-5-amine

A145544-10G A145544-1G A145544-250MG A145544-5G
76997-042EA 588.8 USD
76997-042 76997-044 76997-046 76997-048
2-(4-Methoxy-benzyl)-2H-pyrazol-3-ylamine 95%
2-(4-Methoxy-benzyl)-2H-pyrazol-3-ylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
MDL Number: MFCD03596770
CAS Number: 3528-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR