4-(1H-Purin-6-yl)morpholine 95%

Supplier: Ambeed
Warning

A145525-100MG A145525-1G
77010-498EA 29.71 USD
77010-498 77010-500
4-(1H-Purin-6-yl)morpholine 95%
4-(1H-Purin-6-yl)morpholine

Precaución: for Research Use Only

Formula: C₉H₁₁N₅O
MW: 205.21 g/mol
MDL Number: MFCD09999140
CAS Number: 2846-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR