5-(4-Bromophenyl)-3-methylisoxazole-4-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A145490-100MG
A145490-1G
A145490-250MG
A145490-5G
76984-434EA
20.16
USD
76984-434
76984-436
76984-438
76984-440
5-(4-Bromophenyl)-3-methylisoxazole-4-carboxylic acid 97%
5-(4-Bromophenyl)-3-methylisoxazole-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₈BrNO₃ MW: 282.1 g/mol |
MDL Number:
MFCD16447083 CAS Number: 91182-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...