2-Bromo-3-octylthiophene 95%

Supplier: Ambeed
Warning

A145148-100G A145148-10G A145148-1G A145148-25G A145148-5G
77027-852EA 247 USD
77027-852 77027-854 77027-856 77027-858 77027-860
2-Bromo-3-octylthiophene 95%
2-Bromo-3-octylthiophene

Precaución: for Research Use Only

Formula: C₁₂H₁₉BrS
MDL Number: MFCD13193238
CAS Number: 145543-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR