3-Aminopyrazine-2-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-aminopyrazine-2-carbaldehyde

A145133-100MG A145133-250MG A145133-5G A145133-1G
77027-846EA 63.66 USD
77027-846 77027-848 77027-850 77667-082
3-Aminopyrazine-2-carbaldehyde 97%
3-Aminopyrazine-2-carbaldehyde

Precaución: for Research Use Only

Formula: C5H5N3O
MW: 123.1100 g/mol
MDL Number: MFCD10696886
CAS Number: 32710-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR