(1H-1,2,4-Triazol-1-yl)methanol 95%

Supplier: Ambeed
Warning

A145118-1G A145118-5G
76984-426EA 6.37 USD
76984-426 77683-246
(1H-1,2,4-Triazol-1-yl)methanol 95%
(1H-1,2,4-Triazol-1-yl)methanol

Precaución: For research use only.

Formula: C₃H₅N₃O
MW: 99.09 g/mol
MDL Number: MFCD00227137
CAS Number: 74205-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR