3-Nitro-4-(1-pyrazolyl)benzoic Acid 97%

Supplier: Ambeed
Warning

A145088-10G A145088-1G A145088-5G
77027-840EA 204.76 USD
77027-840 77027-842 77027-844
3-Nitro-4-(1-pyrazolyl)benzoic Acid 97%
3-Nitro-4-(1-pyrazolyl)benzoic Acid

Precaución: for Research Use Only

Formula: C₁₀H₇N₃O₄
MW: 233.18 g/mol
MDL Number: MFCD00141982
CAS Number: 162848-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR