(4-Chloropyrazol-1-yl)acetic acid 97%

Supplier: Ambeed
Warning

A145085-1G A145085-250MG
77027-836EA 28 USD
77027-836 77027-838
(4-Chloropyrazol-1-yl)acetic acid 97%
(4-Chloropyrazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MDL Number: MFCD00036053
CAS Number: 32089-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR