N,N,N',N'-Tetrakis(4-biphenylyl)benzidine 97%

Supplier: Ambeed
Warning

Synonyms: N4,N4,N4',N4'-Tetra([1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4,4'-diamine

A145014-10G A145014-1G A145014-250MG A145014-5G
76984-418EA 207 USD
76984-418 76984-420 76984-422 76984-424
N,N,N',N'-Tetrakis(4-biphenylyl)benzidine 97%
N,N,N',N'-Tetrakis(4-biphenylyl)benzidine

Precaución: for Research Use Only

Formula: C₆₀H₄₄N₂
MW: 793.03 g/mol
MDL Number: MFCD11977303
CAS Number: 164724-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR