(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Tyr(Et)-OH

A144769-10G A144769-1G A144769-250MG A144769-25G A144769-5G
77027-816EA 59.42 USD
77027-816 77027-818 77027-820 77027-822 77027-824
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₅
MW: 309.3575 g/mol
MDL Number: MFCD00065601
CAS Number: 76757-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR