3-Azabicyclo[3.1.0]hexane hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-azabicyclo[3.1.0]hexane hydrochloride

A144626-100MG A144626-1G A144626-250MG A144626-25G A144626-5G
77027-798EA 13.8 USD
77027-798 77027-800 77027-802 77027-804 77027-806
3-Azabicyclo[3.1.0]hexane hydrochloride 97%
3-Azabicyclo[3.1.0]hexane hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉N·ClH
MW: 119.593 g/mol
MDL Number: MFCD12197361
CAS Number: 73799-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR