6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole 95%

Supplier: Ambeed
Warning

A144577-100G A144577-100MG A144577-1G A144577-250MG A144577-5G A144577-25G
77027-788EA 435 USD
77027-788 77027-790 77027-792 77027-794 77027-796 77871-704
6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole 95%
6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFN₂
MDL Number: MFCD16660227
CAS Number: 1231930-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR