Methyl 5,6-dihydroxy-2-methylpyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

A144548-100MG A144548-250MG
76984-384EA 109.28 USD
76984-384 76984-386
Methyl 5,6-dihydroxy-2-methylpyrimidine-4-carboxylate 95%
Methyl 5,6-dihydroxy-2-methylpyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
MDL Number: MFCD09701373
CAS Number: 878158-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR