5,6-Dimethylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A144448-100MG A144448-1G
77010-490EA 20.16 USD
77010-490 77010-492
5,6-Dimethylpyrimidin-4-amine 95%
5,6-Dimethylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.16 g/mol
MDL Number: MFCD09909808
CAS Number: 54568-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR