(10-([1,1'-Biphenyl]-4-yl)anthracen-9-yl)boronic acid 97%

Supplier: Ambeed
Warning

A144429-1G A144429-250MG
77010-482EA 49.87 USD
77010-482 77010-484
(10-([1,1'-Biphenyl]-4-yl)anthracen-9-yl)boronic acid 97%
(10-([1,1'-Biphenyl]-4-yl)anthracen-9-yl)boronic acid

Precaución: for Research Use Only

Formula: C₂₆H₁₉BO₂
MW: 374.24 g/mol
MDL Number: MFCD27955786
CAS Number: 400607-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR