2-(4-Bromopyridin-2-yl)propan-2-ol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromo-2-pyridinyl)-2-propanol

A144423-100MG A144423-1G A144423-250MG
76984-376EA 53.05 USD
76984-376 76984-378 76984-380
2-(4-Bromopyridin-2-yl)propan-2-ol 98%
2-(4-Bromopyridin-2-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
MDL Number: MFCD13190619
CAS Number: 477252-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR